Skip to Content
Merck
All Photos(1)

Key Documents

WHA3001861

Whatman® cellulose chromatography papers

1 Chr sheets, 20 x 20 cm, 100/pk

Synonym(s):

Whatman paper, Z270814, blotting papers, chromatography papers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115727
NACRES:
NB.24
Pricing and availability is not currently available.

material

cellulose membrane
sheets

grade

for chromatography

sterility

non-sterile

feature

1 CHR

packaging

pack of 100

manufacturer/tradename

Whatman 3001-861
Whatman Article No. 28419181 (US reference)

parameter

130 mm/30 min flow rate

W × L

20 cm × 20 cm

basis weight

87 g/m2

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

General description

1 Chr cellulose chromatography roll is a 0.18 mm paper for general analytical separations. The world standard chromatography paper. A smooth surface, 0.18 mm thick with a linear flow rate (water) of 130 mm/30 min. Good resolution for general analytical separations.

Features and Benefits

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service