Skip to Content
Merck
All Photos(1)

Key Documents

W478100

Sigma-Aldrich

L-Cysteine methyl ester hydrochloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
171.65
FEMA Number:
4781
Beilstein:
3685824
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
odorless
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥95%

mp

142 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

Cl[H].COC(=O)[C@@H](N)CS

InChI

1S/C4H9NO2S.ClH/c1-7-4(6)3(5)2-8;/h3,8H,2,5H2,1H3;1H/t3-;/m0./s1

InChI key

WHOHXJZQBJXAKL-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service