Skip to Content
Merck
All Photos(1)

Key Documents

W419001

Sigma-Aldrich

L-Ornithine hydrochloride

99%

Synonym(s):

(S)-(+)-2,5-Diaminopentanoic acid hydrochloride, (S)-2,5-Diaminopentanoic acid monohydrochloride, L-Ornithine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3CH(CO2H)(NH2) · HCl
CAS Number:
Molecular Weight:
168.62
FEMA Number:
4190
Beilstein:
3625847
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

99%

mp

245 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Subchronic tolerance trials of graded oral supplementation with ornithine hydrochloride or citrulline in healthy adults.: This research examines the safety and tolerance of varying doses of L-ornithine hydrochloride in healthy adults. The findings indicate that L-ornithine hydrochloride is well-tolerated at different dosage levels, making it a viable supplement for various applications (Miura et al., 2023).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming
Charles D Mills
Critical reviews in immunology, 32(6), 463-488 (2013-02-23)
The purpose of immunology is simple. Cure or prevent disease. M1/M2 is useful because it is simple. M1/M2 describes the two major and opposing activities of macrophages. M1 activity inhibits cell proliferation and causes tissue damage while M2 activity promotes
Takuya Ouchi et al.
Nature chemical biology, 9(4), 277-283 (2013-02-26)
LysW has been identified as a carrier protein in the lysine biosynthetic pathway that is active through the conversion of α-aminoadipate (AAA) to lysine. In this study, we found that the hyperthermophilic archaeon, Sulfolobus acidocaldarius, not only biosynthesizes lysine through
Janet Newman et al.
PloS one, 8(3), e58298-e58298 (2013-03-14)
Escherichia coli possesses two acyl ornithine aminotransferases, one catabolic (AstC) and the other anabolic (ArgD), that participate in L-arginine metabolism. Although only 58% identical, the enzymes have been shown to be functionally interchangeable. Here we have purified AstC and have
Nicole Berthold et al.
Antimicrobial agents and chemotherapy, 57(1), 402-409 (2012-11-02)
Proline-rich antimicrobial peptides (PrAMPs) from insects and mammals have recently been evaluated for their pharmaceutical potential in treating systemic bacterial infections. Besides the native peptides, several shortened, modified, or even artificial sequences were highly effective in different murine infection models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service