Merck
  • Interorgan metabolism of ornithine phenylacetate (OP)--a novel strategy for treatment of hyperammonemia.
Interorgan metabolism of ornithine phenylacetate (OP)--a novel strategy for treatment of hyperammonemia.
Biochemical pharmacology (2012-10-30)
Sherry Dadsetan, Michael Sørensen, Lasse K Bak, Hendrik Vilstrup, Peter Ott, Arne Schousboe, Rajiv Jalan, Susanne Keiding, Helle S Waagepetersen
ABSTRACT
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming phenylacetylglutamine (PAGN) which is excreted in urine. The aim of the present study was to elucidate the interorgan metabolism of ornithine and ammonia in cirrhotic rats treated with OP in order to obtain an understanding of the underlying mechanisms of the beneficial effect of the treatment, which are largely unknown. Bile duct ligated cirrhotic rats and SHAM rats were treated with OP or saline for five days. [2,5-(15)N]Ornithine or (15)NH(4)(+) were administered intravenously and the incorporation of (15)N in amino acids as well as the content of the amino acids were subsequently determined in plasma, skeletal muscle, liver and kidney. In BDL rats, OP treatment reduced arterial ammonia concentration and increased that of glutamine 30 min after the treatment but not after 15 h. OP treatment did not increase (15)N labeling in glutamine from [2,5-(15)N]ornithine and (15)NH(4)(+) in skeletal muscle or liver. However, the extent of glutamine labeling from [2,5-(15)N]ornithine or (15)NH(4)(+) was similar in arterial blood and liver and higher than that in skeletal muscle. These findings suggest that the effect of OP was related to hepatic metabolism of ornithine. PAGN could not be detected in urine or blood in any of the rats which may explain why OP treatment only reduced arterial ammonia transiently.
Sigma-Aldrich
L-Ornithine hydrochloride, 99%
Sigma-Aldrich
L-Ornithine monohydrochloride, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioXtra, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
D-Ornithine monohydrochloride, ~98%
Sigma-Aldrich
DL-Ornithine monohydrochloride, ≥99.0% (AT)