Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Ornithine hydrochloride
Synonym(s):
(
S
)-(+)-2,5-Diaminopentanoic acid hydrochloride, (
S
)-2,5-Diaminopentanoic acid monohydrochloride, L-Ornithine monohydrochloride
Linear Formula:
NH
2
(CH
2
)
3
CH(CO
2
H)(NH
2
) · HCl
CAS Number:
3184-13-2
Molecular Weight:
168.62
EC Number:
221-678-6
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Optical Isomer
Organoleptic Class
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
W419001
L-Ornithine hydrochloride,
99%
Expand
View Pricing