Skip to Content
Merck
All Photos(1)

Key Documents

W335207

Sigma-Aldrich

4-Hexen-3-one

≥92%

Synonym(s):

4-Hexen-3-one, predominantly trans

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOC2H5
CAS Number:
Molecular Weight:
98.14
FEMA Number:
3352
EC Number:
Council of Europe no.:
718
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.048
NACRES:
NA.21
grade:
Halal
biological source:
synthetic

biological source

synthetic

grade

Halal

Assay

≥92%

refractive index

n20/D 1.44 (lit.)

bp

135-137 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

Organoleptic

pungent; ethereal

SMILES string

CCC(=O)\C=C\C

InChI

1S/C6H10O/c1-3-5-6(7)4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

FEWIGMWODIRUJM-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hexen-3-one is an unsaturated aldehyde that is reported to occur in strawberry fruit, essential oil of Ruta graveolens and Holigarna grahamii leaves. Due to its fruity fragrance, it is also used in the perfume industry.

Application


  • Key lime (Citrus aurantifolia) inhibits the growth of triple drug resistant Helicobacter pylori.: Explores the antibacterial properties of key lime, potentially including volatile components like 4-Hexen-3-one, highlighting its relevance in medical microbiology and the development of alternative therapeutic agents (Lee et al., 2018).

  • Chemical composition of hexane extract of Citrus aurantifolia and anti-Mycobacterium tuberculosis activity of some of its constituents.: Investigates the components of Citrus aurantifolia hexane extract, including the role of 4-Hexen-3-one in combating Mycobacterium tuberculosis, demonstrating the potential of natural products in treating infectious diseases (Sandoval-Montemayor et al., 2012).

  • Kinetic investigation of the OH radical and Cl atom initiated degradation of unsaturated ketones at atmospheric pressure and 298 K.: This study examines the reactivity of atmospheric ketones including 4-Hexen-3-one with OH radicals and Cl atoms, providing valuable data for understanding their fate in the atmosphere (Blanco et al., 2012).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of Inorganic and Biological Fertilizer Treatments on Essential Oil Composition of Ruta graveolens L.
Malik AA, et al.
Journal of herbs, spices & medicinal plants, 18(2), 191-202 (2012)
Su-Mi Lee et al.
Gut pathogens, 10, 16-16 (2018-06-27)
Eradication rate for Helicobacter pylori (H. pylori) has decreased due to antibiotic resistance. Therefore, new strategies are needed to enhance H. pylori eradication, especially for H. pylori with high antibiotic resistance. The objective of this study was to evaluate anti-H.
GC-MS analysis of bioactive compounds in methanolic extract of Holigarna grahamii (wight) Kurz.
Jadhav V, et al.
Journal of herbs, spices & medicinal plants, 2, 35-39 (2014)
Metabolism of natural volatile compounds by strawberry fruit.
Hamilton-Kemp TR, et al.
Journal of Agricultural and Food Chemistry, 44(9), 2802-2805 (1996)
Guangwei Si et al.
Neuron, 101(5), 950-962 (2019-01-27)
Odor perception allows animals to distinguish odors, recognize the same odor across concentrations, and determine concentration changes. How the activity patterns of primary olfactory receptor neurons (ORNs), at the individual and population levels, facilitate distinguishing these functions remains poorly understood. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service