Skip to Content
Merck
All Photos(1)

Key Documents

W293407

Sigma-Aldrich

Propyl butyrate

≥98%

Synonym(s):

n-Propyl-n-butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH2CH2CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2934
Beilstein:
1745552
EC Number:
Council of Europe no.:
266
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.040
NACRES:
NA.21
Organoleptic:
ethereal; fruity; sweet
grade:
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

Assay

≥98%

refractive index

n20/D 1.4 (lit.)

bp

142-143 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; fruity; sweet

SMILES string

CCCOC(=O)CCC

InChI

1S/C7H14O2/c1-3-5-7(8)9-6-4-2/h3-6H2,1-2H3

InChI key

HUAZGNHGCJGYNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural Occurrence: Apple, banana, kiwi, melon, papaya, passion fruit, rum and strawberry.

Biochem/physiol Actions

Taste at 20 ppm

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 67-67 (2005)
Jorge Montesinos et al.
Brain, behavior, and immunity, 45, 233-244 (2014-12-09)
The adolescent brain undergoes important dynamic and plastic cell changes, including overproduction of axons and synapses, followed by rapid pruning along with ongoing axon myelination. These developmental changes make the adolescent brain particularly vulnerable to neurotoxic and behavioral effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service