Skip to Content
Merck
All Photos(1)

Key Documents

W329304

Sigma-Aldrich

α-Angelica lactone

98%, FG

Synonym(s):

4-Hydroxy-3-pentenoic acid γ-lactone, 5-Methyl-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
FEMA Number:
3293
Beilstein:
108394
EC Number:
Council of Europe no.:
731
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.012
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

98%

refractive index

n20/D 1.448 (lit.)

bp

55-56 °C/12 mmHg (lit.)

mp

13-17 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; oily; sweet

SMILES string

CC1=CCC(=O)O1

InChI

1S/C5H6O2/c1-4-2-3-5(6)7-4/h2H,3H2,1H3

InChI key

QOTQFLOTGBBMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α-Angelica lactone, a cyclic lactone that occurs naturally in tobacco, is generally used in tobacco flavoring.

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition and carcinogenicity of smokeless tobacco.
Hoffmann D & Djordjevic MV.
Advances in Dental Research, 11(3), 322-329 (1997)
Efficient conversion of ?-angelica lactone into ?-valerolactone with ionic liquids at room temperature
Cao R, et al.
ACS sustainable chemistry & engineering, 2(4), 902-909 (2014)
W A Nijhoff et al.
Carcinogenesis, 16(3), 607-612 (1995-03-01)
The naturally occurring anticarcinogens flavone and alpha-angelicalactone incorporated separately and simultaneously in the diet at 0.5, 0.1, 0.05 and 0.01% w/w, were studied with respect to their effects on oesophageal, gastric, intestinal, colonic and hepatic (i) glutathione S-transferase (GST) enzyme
Y M Ioannou et al.
Cancer research, 42(4), 1199-1204 (1982-04-01)
The effects of alpha-angelica lactone (alpha-AL), butylated hydroxyanisole (BHA), and beta-naphthoflavone (beta-NF) on the amount of benzo(alpha)pyrene (BP) metabolite:DNA adducts formed in the forestomach, lung, and liver of ICR/Ha mice were investigated 48 hr after p.o. administration of BP. BP
Lin Zhou et al.
Organic letters, 13(12), 3056-3059 (2011-05-21)
A direct highly diastereo- and enantioselective asymmetric vinylogous Mannich-type (AVM) reaction of aldimines with nonactivated natural α-angelica lactone has been successfully developed. It was demonstrated that the nonactivated natural α-angelica lactone is a useful vinylogous nucleophile to give the chiral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service