Skip to Content
Merck
All Photos(1)

Key Documents

W310301

Sigma-Aldrich

γ-Valerolactone

≥99%, FCC, FG

Synonym(s):

γ-Methyl-γ-butyrolactone, 4,5-Dihydro-5-methyl-2(3H)-furanone, 4-Hydroxypentanoic acid lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
FEMA Number:
3103
Beilstein:
80420
EC Number:
Council of Europe no.:
757
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.013
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

vapor density

3.45 (vs air)

Assay

≥99%

refractive index

n20/D 1.432 (lit.)

bp

207-208 °C (lit.)
82-85 °C/10 mmHg (lit.)

mp

−31 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cocoa; herbaceous; woody; sweet; warm

SMILES string

CC1CCC(=O)O1

InChI

1S/C5H8O2/c1-4-2-3-5(6)7-4/h4H,2-3H2,1H3

InChI key

GAEKPEKOJKCEMS-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

γ-Valerolactone has been identified as one of the volatile flavor constituents in mango and honey.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Importance of some lactones and 2, 5-dimethyl-4-hydroxy-3 (2H)-furanone to mango (Mangifera indica L.) aroma.
Wilson III CW, et al.
Journal of Agricultural and Food Chemistry, 38(7), 1556-1559 (1990)
S Hein et al.
FEMS microbiology letters, 153(2), 411-418 (1997-08-15)
The aim of this study was the production of the homopolyester poly(4-hydroxybutyric acid) (poly(4HB)) with recombinant strains of Escherichia coli. Wild-type strains and other widely used non-recombinant strains of E. coli are not able to produce polyhydroxyalkanoic acids (PHA) as
Floral origin markers of heather honeys: Erica arborea and Calluna vulgaris.
Guyot C, et al.
Food Chemistry, 64(1), 3-11 (1999)
Conversion of hemicellulose into furfural using solid acid catalysts in γ-valerolactone.
Elif I Gürbüz et al.
Angewandte Chemie (International ed. in English), 52(4), 1270-1274 (2012-12-06)
M Iwata et al.
International archives of occupational and environmental health, 51(3), 253-260 (1983-01-01)
n-Hexane is one of the solvents widely used in industry and well known to be neurotoxic. Recently it was clearly revealed that n-hexane is metabolized in vivo and its metabolites are excreted in the urine. However, the relationship between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service