Skip to Content
Merck
All Photos(1)

Key Documents

W236101

Sigma-Aldrich

δ-Decalactone

≥98%, FCC, FG

Synonym(s):

(±)-5-Decanolide, δ-Decanolactone, (±)-δ-Pentyl-δ-valerolactone, (±)-6-Pentyltetrahydro-2H-pyran-2-one, 5-Hydroxydecanoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2361
Beilstein:
117520
EC Number:
Council of Europe no.:
621
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.007
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.458 (lit.)

bp

117-120 °C/0.02 mmHg (lit.)

mp

−27 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; creamy; fatty; milk; peach; sweet

SMILES string

CCCCCC1CCCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

GHBSPIPJMLAMEP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

δ-Decalactone is the main flavor compound found in nonfat dry milk, evaporated and dried whole milk. It is also reported to be responsible for the coconut-like off-flavor in fat-containing dairy products.

Other Notes

Natural occurrence: Sherry, white wine, mango, cassava, loganberry and fresh plum.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 21, 51-51 (2002)
Aroma-active components of nonfat dry milk
Karagul-Yuceer Y, et al.
Journal of Agricultural and Food Chemistry, 49(6), 2948-2953 (2001)
Gas Chromatographic Analysis of Flavor in Processed Milks1, 2.
Patton S.
The Journal of Dairy Research, 44(2), 207-214 (1961)
The Coconut-Like Flavor Defect of Milk Fat. II. Demonstration of ?-Decalactone in Dried Cream, Dry Whole Milk, and Evaporated Milk1.
Keeney PG & Patton S.
The Journal of Dairy Research, 39(8), 1114-1119 (1956)
Pascal Fuchsmann et al.
Journal of chromatography. A, 1601, 60-70 (2019-06-11)
Headspace in-tube extraction (HS-ITEX) and solid phase microextraction (HS-SPME) sampling, followed by gas chromatography-mass spectrometry (GC-MS), are widely used to analyze volatile compounds in various food matrices. While the extraction efficiency of volatile compounds from foodstuffs is crucial for obtaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service