Skip to Content
Merck
All Photos(1)

Key Documents

W314900

Sigma-Aldrich

2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers

≥95%, FG

Synonym(s):

2-ethyl-3,5(or 6)-dimethylpyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3149
Council of Europe no.:
2246
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.100
NACRES:
NA.21
Organoleptic:
chocolate; hazelnut; nutty; peanut; roasted
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥95%

refractive index

n20/D 1.5015 (lit.)

bp

180-181 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

chocolate; hazelnut; nutty; peanut; roasted

SMILES string

CCc1ncc(C)nc1C.CCc2nc(C)cnc2C

InChI

1S/2C8H12N2/c1-4-8-7(3)10-6(2)5-9-8;1-4-8-7(3)9-5-6(2)10-8/h2*5H,4H2,1-3H3

InChI key

BOFLOMVCGPWPQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Ethyl-3(5 or 6)-dimethylpyrazine, a mixture of isomers are odor-active pyrazine compounds that are typically found in roasted foods formed due to Maillard reaction and pyrolysis of serine and threonine.

Application


  • Characterization of Flavor Profile of Sauced Pork from Different Regions of China Based on E-Nose, E-Tongue and Gas Chromatography-Ion Mobility Spectroscopy.: This study by Yuan et al. characterizes the flavor profile of sauced pork using various sensory and chromatographic techniques, including gas chromatography-ion mobility spectroscopy. It identifies key aroma compounds, including 2-ethyl-3,5(6)-dimethylpyrazine, contributing to the overall flavor profile (Yuan et al., 2024).

  • The effect of thermal times of circulating non-fried roast technique on the formation of (non)volatile compounds in roasted mutton by multi-chromatography techniques and heat transfer analysis.: Liu et al. explore how different thermal processing times affect the formation of volatile and non-volatile compounds in roasted mutton, highlighting the presence of 2-ethyl-3,5(6)-dimethylpyrazine among the key aroma contributors (Liu et al., 2023).

  • Investigation of the effect of over-fired drying on the taste and aroma of Lu′an Guapian tea using metabolomics and sensory histology techniques.: Zhang et al. investigate how over-fired drying affects the aroma and taste of Lu′an Guapian tea, identifying 2-ethyl-3,5(6)-dimethylpyrazine as a significant aroma compound (Zhang et al., 2024).

  • Unveiling the aromatic intricacies of Wuyi Rock Tea: A comparative study on sensory attributes and odor-active compounds of Rougui and Shuixian varieties.: This comparative study by Wang et al. examines the sensory attributes and odor-active compounds of Wuyi Rock Tea, highlighting the role of 2-ethyl-3,5(6)-dimethylpyrazine in the aroma profile (Wang et al., 2024).

  • Characterization of the Key Aroma Compounds in Dong Ding Oolong Tea by Application of the Sensomics Approach.: Wang et al. characterize the key aroma compounds in Dong Ding Oolong tea, identifying 2-ethyl-3,5(6)-dimethylpyrazine as one of the major contributors to its distinct aroma profile (Wang et al., 2023).

Biochem/physiol Actions

Taste at 5 ppm

Other Notes

Natural occurrence: Burley and Virginia tobacco, cocoa, coffee, cooked beef, peanut, Swiss cheese.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F

Flash Point(C)

69 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Model reactions on roast aroma formation.
Baltes W & Bochmann G.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 184(6), 485-493 (1987)
Mosciano, G.
Perfumer & Flavorist, 30, 57-57 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 41-41 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service