Skip to Content
Merck
All Photos(3)

Key Documents

W249311

Sigma-Aldrich

Furfuryl mercaptan

greener alternative

natural, 98%, FG

Synonym(s):

2-Furanmethanethiol, 2-Furfurylthiol, 2-Furylmethanethiol, Furfuryl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
FEMA Number:
2493
Beilstein:
383594
EC Number:
Council of Europe no.:
2202
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.026
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.531 (lit.)

bp

155 °C (lit.)

density

1.132 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

coffee; meaty; roasted; sulfurous

SMILES string

SCc1ccco1

InChI

1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2

InChI key

ZFFTZDQKIXPDAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Other Notes

Natural occurrence: Burley tobacco, bread, cocoa, coffee, juniper berry oil, potato chips, roasted barley, whiskey.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imre Blank et al.
Journal of agricultural and food chemistry, 50(8), 2356-2364 (2002-04-04)
The stability of the coffee flavor compound furfuryl mercaptan has been investigated in aqueous solutions under Fenton-type reaction conditions. The impact of hydrogen peroxide, iron, ascorbic acid, and ethylenediaminetetraacetic acid was studied in various combinations of reagents and temperature. Furfuryl
Luigi Poisson et al.
Journal of agricultural and food chemistry, 57(21), 9923-9931 (2009-10-13)
The formation of several key odorants, such as 2-furfurylthiol (FFT), alkylpyrazines, and diketones, was studied upon coffee roasting. The approach involved the incorporation of potential precursors in green coffee beans by means of biomimetic in-bean and spiking experiments. Both labeled
Brian G Lake et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(12), 1761-1770 (2003-10-18)
The metabolism of two thiofurans, namely furfuryl mercaptan (FM) and 2-methyl-3-furanthiol (MTF), to their corresponding methyl sulphide and methyl sulphoxide derivatives has been studied in male Sprague-Dawley rat hepatocytes and liver microsomes. Rat hepatocytes converted FM to furfuryl methyl sulphoxide
Christoph Müller et al.
Journal of agricultural and food chemistry, 53(7), 2623-2629 (2005-03-31)
The purpose of the following study was to investigate the influence of coffee roasting on the thiol-binding activity of coffee beverages, and to investigate the potential of various green bean compounds as precursors of thiol-binding sites by using promising "in
Segolene Leclercq et al.
Journal of agricultural and food chemistry, 55(22), 9189-9194 (2007-10-12)
The objective of this study was to investigate approaches to protect selected flavor compounds from deterioration when stored in an oil matrix. An aroma compound model mixture was prepared in a medium-chain triglyceride (MCT) or sunflower oil (SfO) matrix and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service