W214701
Benzyl mercaptan
99%, FG
Synonym(s):
α-Toluenethiol, α-Tolyl mercaptan, Benzenemethanethiol
About This Item
biological source
synthetic
Quality Level
grade
FG
Kosher
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515
vapor density
>4 (vs air)
Assay
99%
impurities
≤0.0890% Toluene, residual
refractive index
n20/D 1.575 (lit.)
bp
194-195 °C (lit.)
density
1.058 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
garlic; onion; alliaceous; sulfurous
SMILES string
SCc1ccccc1
InChI
1S/C7H8S/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
InChI key
UENWRTRMUIOCKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Anaerobic degradation of thiobencarb by mixed culture of isolated bacteria.: This research focuses on the anaerobic degradation of thiobencarb, a thiol-related compound, by a mixed culture of bacteria, highlighting potential bioremediation applications (Duc et al., 2023).
- Amphiphilic polymer-encapsulated Au nanoclusters with enhanced emission and stability for highly selective detection of hypochlorous acid.: This study demonstrates the use of amphiphilic polymer-encapsulated gold nanoclusters for the selective detection of hypochlorous acid, indicating potential applications in environmental monitoring and chemical sensing (Li et al., 2021).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
158.0 °F - closed cup
Flash Point(C)
70 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service