Merck
  • Effect of type of oil and addition of delta-tocopherol on model flavor compound stability during storage.
Effect of type of oil and addition of delta-tocopherol on model flavor compound stability during storage.
Journal of agricultural and food chemistry (2007-10-12)
Segolene Leclercq, Gary A Reineccius, Christian Milo
ABSTRACT
The objective of this study was to investigate approaches to protect selected flavor compounds from deterioration when stored in an oil matrix. An aroma compound model mixture was prepared in a medium-chain triglyceride (MCT) or sunflower oil (SfO) matrix and stored under either an ambient air or argon atmosphere containing, respectively, ca. 20 and <0.5% residual oxygen as controls or containing a natural antioxidant, delta-tocopherol (0.01%). Samples were analyzed by static headspace GC/FID to determine the stability over time of the compounds in mixture. It was found that the type of oil had the greatest effect (P < 0.01) on overall compound stability. A low-oxygen atmosphere also had a significant (P < 0.05) protective effect on the aroma compounds in both oils. The addition of delta-tocopherol generally offered little additional protection. No significant relationship could be determined between the oxidation of the lipid matrix and the loss of oxidation-sensitive thiol compounds.
Sigma-Aldrich
2-Furanmethanethiol, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Methylpyrrole, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Furfuryl mercaptan, ≥97%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Furfuryl mercaptan, natural, 98%, FG