Skip to Content
Merck
All Photos(1)

Key Documents

W242705

Sigma-Aldrich

Ethyl butyrate

≥98%, FCC, FG

Synonym(s):

Butyric acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(O)OC2H5
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2427
Beilstein:
506331
EC Number:
Council of Europe no.:
264
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.039
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

4 (vs air)

vapor pressure

15.5 mmHg ( 25 °C)

Assay

≥98%

autoignition temp.

865 °F

refractive index

n20/D 1.392 (lit.)

bp

120 °C (lit.)

mp

−93 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; fruity; pineapple; sweet

SMILES string

CCCC(=O)OCC

InChI

1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3

InChI key

OBNCKNCVKJNDBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Timolol transport from microemulsions trapped in HEMA gels.: This article examines the microemulsion systems for drug delivery, where ethyl butyrate could play a role in solubilizing components (Li CC et al., 2007).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Cerf-Ducastel et al.
Physiology & behavior, 81(3), 389-396 (2004-05-12)
A protocol suited to the investigations of odor-taste interactions in a functional magnetic resonance imaging (fMRI) scanner is described and results that confirm the validity of that protocol are presented. Stimuli were dissolved in water and presented to the mouth
Montserrat Mestres et al.
Journal of agricultural and food chemistry, 54(5), 1814-1821 (2006-03-02)
The influence of gel texture on parameters such as positioning of food material in the oral cavity during mastication, and salivation, and their influence on aroma release in vivo was studied. Retronasal perception was followed by means of time-resolved sensory
Geraldine Savary et al.
Journal of agricultural and food chemistry, 54(3), 665-671 (2006-02-02)
Diffusion-ordered spectroscopy (DOSY)-pulsed field gradient (PGF) nuclear magnetic resonance (NMR) spectroscopy was used to measure self-diffusion coefficients of aroma molecules in model fruit preparations. The impact of the sucrose content on aroma diffusion was specifically investigated, and the relationship with
Marcos Pita et al.
Physical chemistry chemical physics : PCCP, 13(10), 4507-4513 (2011-01-25)
We realize a biochemical filtering process by introducing a buffer in a biocatalytic signal-transduction logic system based on the function of an enzyme, esterase. The input, ethyl butyrate, is converted into butyric acid--the output signal, which in turn is measured
Penelope A Lind et al.
Biochimica et biophysica acta, 1702(1), 103-110 (2004-09-29)
It has been generally accepted that enzyme activity requires a minimal hydration of about 0.2 g H2O g(-1) protein. This fits well with evidence that hydration above this level is associated with the onset of intramolecular motions. The influence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service