Skip to Content
Merck
All Photos(1)

Key Documents

W213500

Sigma-Aldrich

Benzyl acetate

≥99%, FCC, FG

Synonym(s):

Benzyl ethanoate, Acetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2135
Beilstein:
1908121
EC Number:
Council of Europe no.:
204
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.014
NACRES:
NA.21
Organoleptic:
floral; fruity; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

23 mmHg ( 110 °C)

Assay

≥99%

autoignition temp.

862 °F

refractive index

n20/D 1.502 (lit.)

bp

206 °C (lit.)

mp

−51 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; fruity; sweet

SMILES string

CC(=O)OCc1ccccc1

InChI

1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QUKGYYKBILRGFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzyl acetate is an aliphatic flavor ester found in the oils of many plants such as jasmine, hyacinth, gardenias, and azaleas.[1][2]

Application


  • Transcriptomic and proteomic approaches to explore the differences in monoterpene and benzenoid biosynthesis between scented and unscented genotypes of wintersweet.: This study examines the biosynthesis pathways of monoterpenes and benzenoids, including benzyl acetate, in different genotypes of wintersweet. By using transcriptomic and proteomic methods, the researchers identified key genes and proteins involved in the production of these compounds, providing insights into the biochemical mechanisms underlying their biosynthesis. This research has implications for understanding the metabolic engineering of fragrance compounds in plants (Tian et al., 2019).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 260-261 (1997)
Modeling, Simulation, and Kinetic Studies of Solvent-Free Biosynthesis of Benzyl Acetate
Garlapati VK, et al
Journal of Chemistry (2013)
G Y Wittman et al.
Journal of chromatography. A, 874(2), 225-234 (2000-05-19)
The direct derivatisation of acetic acid with n-hexyl chloroformate and with benzyl bromide in water was evaluated. With n-hexyl chloroformate, acetic acid did not give the n-hexyl acetate derivative, but the reaction of acetic acid with benzyl bromide in aqueous
Sotaro Momosaki et al.
Nuclear medicine and biology, 34(8), 939-944 (2007-11-14)
In order to develop a suitable radiotracer for the measurement of glial metabolism, we synthesized four different types of ester derivatives of [14C]acetate, namely, [14C]phenyl acetate, [14C]para-nitrophenyl acetate, [14C]2,4-dinitrophenyl acetate and [14C]benzyl acetate ([14C]BA), and evaluated their potencies in rats.
J Caldwell
Teratogenesis, carcinogenesis, and mutagenesis, 13(4), 185-190 (1993-01-01)
The mouse lymphoma assay is commonly included in batteries of tests for the mutagenic and carcinogenic potential of chemicals. Unlike other bacterial and mammalian cell tests, it has an unacceptably high incidence of false positive results. This paper reviews data

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service