Skip to Content
Merck
All Photos(1)

Key Documents

W241407

Sigma-Aldrich

Ethyl acetate

≥99%, FCC, FG

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2414
Beilstein:
506104
EC Number:
Council of Europe no.:
191
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.001
NACRES:
NA.01

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.560
FDA 21 CFR 182.60

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

format

neat

Organoleptic

green; ethereal; fruity; sweet

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate (EtOAc) is an effective alternate solvent for diethyl ether, employed for the concentration of eggs, larvae, and cysts in fecal specimens during the Formalin-ether sedimentation technique. Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of Kober chromogen into tetrachloroethane containing trichloroacetic acid in determination of urinary estrogens during pregnancy.
N Y Yang et al.
Clinical chemistry, 24(12), 2217-2217 (1978-12-01)
Andreas F Thünemann et al.
Langmuir : the ACS journal of surfaces and colloids, 22(5), 2351-2357 (2006-02-24)
Superparamagnetic iron oxide particles (SPIO) of maghemite were prepared in aqueous solution and subsequently stabilized with polymers in two layer-by-layer deposition steps. The first layer around the maghemite core is formed by poly(ethylene imine) (PEI), and the second one is
Complete oxidation of CO, ethanol, and ethyl acetate over copper oxide supported on titania and ceria modified titania.
Larsson PO and Andersson A.
J. Catal., 179(1), 72-89 (1998)
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration
Junwei Liu et al.
Planta, 241(6), 1435-1451 (2015-02-27)
Strigolactone changes and cross talk with ABA unveil a picture of root-specific hormonal dynamics under stress. Strigolactones (SLs) are carotenoid-derived hormones influencing diverse aspects of development and communication with (micro)organisms, and proposed as mediators of environmental stimuli in resource allocation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service