W238902
2,6-Dimethyl-5-heptenal
stabilized, FCC, FG
Synonym(s):
Melonal
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515
description
may contain a suitable antioxidant
Assay
85%
contains
α-tocopherol as stabilizer (synthetic)
refractive index
n20/D 1.444 (lit.)
bp
116-124 °C/100 mmHg (lit.)
density
0.851 g/mL at 25 °C
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
cucumber; green; melon; sweet; watermelon
SMILES string
CC(CC\C=C(\C)C)C=O
InChI
1S/C9H16O/c1-8(2)5-4-6-9(3)7-10/h5,7,9H,4,6H2,1-3H3
InChI key
YGFGZTXGYTUXBA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Food volatile compounds facilitating HII mesophase formation: solubilization and stability.: This 2011 study explores the role of food volatile compounds, including 2,6-dimethyl-5-heptenal, in facilitating the formation and stability of HII mesophase, a structure relevant in food chemistry and material science (Amar-Zrihen et al., 2011).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1B
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
143.6 °F
Flash Point(C)
62 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service