Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,6-Dimethyl-5-heptenal
Synonym(s):
Melonal
Linear Formula:
(CH
3
)
2
C=CHCH
2
CH
2
CH(CH
3
)CHO
CAS Number:
106-72-9
Molecular Weight:
140.22
EC Number:
203-427-2
Products
Product Category
aroma chemicals (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (2)
Fragrance grade (2)
Halal (2)
Kosher (2)
Natural (1)
Technical (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
W238902
2,6-Dimethyl-5-heptenal,
stabilized, FCC, FG
Expand
View Pricing
W238920
2,6-Dimethyl-5-heptenal,
mixture of isomers, natural, FG
Expand
View Pricing
307963
2,6-Dimethyl-5-heptenal,
technical grade, 80%
Expand
View Pricing