Skip to Content
Merck
All Photos(1)

Key Documents

27470

Sigma-Aldrich

(±)-Citronellal

≥95.0% (GC)

Synonym(s):

(±)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
Beilstein:
1720789
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

207 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CC(CC\C=C(\C)C)CC=O

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3

InChI key

NEHNMFOYXAPHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Citronellal was used to inhibit the development of mechanical nociception induced by carrageenan (CG) and tumor necrosis factor-α (TNF-α).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raimundo Wagner de S Aguiar et al.
TheScientificWorldJournal, 2014, 492138-492138 (2014-03-07)
Corymbia citriodora and Cymbopogon nardus essential oils samples were analyzed by GC and GC-MS and their qualitative and quantitative compositions established. The main component of essential oils of C. citriodora and C. nardus was citronellal, at 61.78% and 36.6%, respectively.
Hajo Kries et al.
The Journal of biological chemistry, 292(35), 14659-14667 (2017-07-14)
The natural product class of iridoids, found in various species of flowering plants, harbors astonishing chemical complexity. The discovery of iridoid biosynthetic genes in the medicinal plant Catharanthus roseus has provided insight into the biosynthetic origins of this class of
Md Munir Mostafiz et al.
Insects, 12(1) (2021-01-06)
The Indian meal moth, Plodia interpunctella (Hübner) (Lepidoptera: Pyralidae), is an insect pest that commonly affects stored and postharvest agricultural products. For the control of insect pests and mites, methyl benzoate (MBe) is lethal as a fumigant and also causes
Nadia Melo et al.
Current biology : CB, 30(1), 127-134 (2019-12-17)
Geosmin is one of the most recognizable and common microbial smells on the planet. Some insects, like mosquitoes, require microbial-rich environments for their progeny, whereas for other insects such microbes may prove dangerous. In the vinegar fly Drosophila melanogaster, geosmin
Walter S Leal et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(46), 18704-18709 (2013-10-30)
The southern house mosquito, Culex quinquefasciatus, has one of the most acute and eclectic olfactory systems of all mosquito species hitherto studied. Here, we used Illumina sequencing to identify olfactory genes expressed predominantly in antenna, mosquito's main olfactory organ. Less

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service