Skip to Content
Merck
All Photos(1)

Key Documents

W230715

Sigma-Aldrich

(±)-Citronellal

natural, ≥85%, FCC, FG

Synonym(s):

(±)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2307
Beilstein:
1720789
EC Number:
Council of Europe no.:
110c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.021
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥85%

refractive index

n20/D 1.451 (lit.)

bp

207 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; rose; sweet; citrus

SMILES string

CC(CC\C=C(\C)C)CC=O

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3

InChI key

NEHNMFOYXAPHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heike Demmer et al.
Journal of neurophysiology, 102(3), 1538-1550 (2009-06-26)
The insect mushroom bodies (MBs) are multimodal signal processing centers and are essential for olfactory learning. Electrophysiological recordings from the MBs' principal component neurons, the Kenyon cells (KCs), showed a sparse representation of olfactory signals. It has been proposed that
Walter S Leal et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(46), 18704-18709 (2013-10-30)
The southern house mosquito, Culex quinquefasciatus, has one of the most acute and eclectic olfactory systems of all mosquito species hitherto studied. Here, we used Illumina sequencing to identify olfactory genes expressed predominantly in antenna, mosquito's main olfactory organ. Less
Lucindo Quintans-Júnior et al.
Journal of medicinal food, 14(6), 630-639 (2011-04-13)
Citronellal (CT) is a monoterpenoid and the major constituent of the mixture of terpenoids that give the citronella oil its lemon scent. Citronella oil is widely used around the world for various purposes and is mainly obtained from plants of
M S Melo et al.
Pharmaceutical biology, 48(4), 411-416 (2010-07-22)
Citronellal is a monoterpene reported to be a major component of the essential oils in various aromatic species of plants. The present study evaluated the central nervous system depressant and antinociceptive properties of citronellal through behavioral experimental models. Following intraperitoneal
Marisa Belicchi-Ferrari et al.
Journal of inorganic biochemistry, 104(2), 199-206 (2009-12-08)
This paper reports the syntheses and characterization of ethylmorpholine substituted citronellal thiosemicarbazone copper(II) and nickel(II) metal complexes. The compounds were characterized through elemental analyses and spectroscopic (IR, UV-Vis, NMR, MS) methods. The X-ray analysis of the two complexes shows that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service