Skip to Content
Merck
All Photos(1)

Key Documents

W296309

Sigma-Aldrich

(R)-(+)-Pulegone

≥90%

Synonym(s):

(+)-Pulegone, (R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one, p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
2963
Beilstein:
2040703
EC Number:
Council of Europe no.:
2050c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Kosher

vapor pressure

138 mmHg ( 25 °C)

Assay

≥90%

optical activity

[α]20/D +22°, neat

refractive index

n20/D 1.486 (lit.)

bp

224 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; minty; sulfurous; sweet

SMILES string

C[C@@H]1CC\C(=C(\C)C)C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 20 ppm

Other Notes

Natural occurrence: Buchu oil, native spearmint, pennyroyal, peppermint, scotch spearmint and thyme.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 56-56 (2005)
Mitscheli S Da Rocha et al.
Toxicological sciences : an official journal of the Society of Toxicology, 128(1), 1-8 (2012-04-14)
Essential oils from mint plants, including peppermint and pennyroyal oils, are used at low levels as flavoring agents in various foods and beverages. Pulegone is a component of these oils. In a 2-year bioassay, oral administration of pulegone slightly increased
Chang-Qin Li et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(21), 3173-3175 (2013-02-13)
To study the validity of Schizonepeta tenuifalia pieces. The change principle for the contents of essential oil and pulegone was determined by the way of classical constant temperature acceleration experiment, and the reacting speed at 20 degrees C was calculated
Maung Kyaw Moe Tun et al.
The Journal of organic chemistry, 77(20), 9422-9425 (2012-10-04)
A three-step synthesis of (R)-(+)-4-methylcyclohex-2-ene-1-one (1) from (R)-(+)-pulegone (3), proceeding in 44% overall yield, is described. The sequence comprises vinyl triflate formation, site-selective ozonolysis, and reduction. The route requires only one chromatographic purification and provides a convenient method to access
Charalabos D Kanakis et al.
Phytochemical analysis : PCA, 23(1), 34-43 (2011-05-28)
Mentha pulegium L. (pennyroyal) is one of the four most commercially important Mentha species, even it is not a cultivated plant. It can be abundantly located in the Iberian Peninsula and North African countries. In Greece it grows in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service