W215201
Benzyl isovalerate
≥98%, FCC, FG
Synonym(s):
Benzyl 3-methylbutyrate
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515
Assay
≥98%
composition
contains IFRA restricted Benzyl benzoate and Benzyl alcohol
refractive index
n20/D 1.488 (lit.)
bp
116 °C/9 mmHg (lit.)
density
0.988 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
benzyl alcohol, benzyl benzoate
Organoleptic
fruity; sweet
SMILES string
CC(C)CC(=O)OCc1ccccc1
InChI
1S/C12H16O2/c1-10(2)8-12(13)14-9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
InChI key
HVJKZICIMIWFCP-UHFFFAOYSA-N
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service