Skip to Content
Merck
All Photos(1)

Key Documents

MAK334

Sigma-Aldrich

Antioxidant Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Oxidative Stress Assay Kit, Total Antioxidant Capacity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

usage

sufficient for 100 colorimetric tests

input

biological sample(s)
plasma
food(s)
serum
urine
beverage(s)
saliva

application(s)

cosmetics
food and beverages
pharmaceutical

detection method

colorimetric

relevant disease(s)

Alzheimer′s disease; Parkinson′s disease; diabetes; neurological disorders

General description

An antioxidant is a molecule capable of slowing or preventing the oxidation of other molecules. Antioxidants protect the cells from damages by reactive oxygen species which are produced in oxidation reactions in the cell. Antioxidants can be small molecules such as glutathione, vitamins, or macromolecules such as catalase, glutathione peroxidase. As oxidative stress contributes to the development of many diseases including Alzheimer′s disease, Parkinson′s disease, diabetes, rheumatoid arthritis and neurodegeneration, the use of antioxidants in pharmacology is intensively studied. Antioxidants are also widely used as dietary supplements and in industry as preservatives in food, cosmetics, rubber and gasoline. Simple, direct and high-throughput assays for total antioxidant capacity (TAC) find wide applications in research, food industry and drug discovery.

Application

The Antioxidant Assay Kit may be used for:
  • Cancer Research
  • Diabetes Research
  • Drug Discovery
  • Neurodegenerative Disease Research
  • Oxidative Stress Research

Features and Benefits

Broad Detection Range: Detect antioxidants accurately across a wide range, from 1.5 µM to 1000 µM Trolox equivalents, using only 20 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.





Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a 10 minute room temperature reaction, reducing complexity and saving valuable time and effort.



Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Compatible with high-throughput handling systems.

Suitability

This kit is suitable for antioxidant determination in serum, plasma, urine, saliva and other biological samples, food and beverages. The kit can also be utilized in drug discovery/pharmacology for studying the effects of drugs on TAC.

Principle

The Antioxidant Assay Kit′s improved assay measures total antioxidant capacity in which Cu2+ is reduced by antioxidant to Cu+. The resulting Cu+ specifically forms a colored complex with a dye reagent. The color intensity at 570nm is proportional to TAC in the sample. The kit uses 20 μL of sample and has a linear detection range from 1.5 to 1000 μM Trolox equivalents.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miklós Pogány et al.
Plants (Basel, Switzerland), 11(7) (2022-04-13)
Noble rot is a favorable form of the interaction between grape (Vitis spp.) berries and the phytopathogenic fungus Botrytis cinerea. The transcriptome pattern of grapevine cells subject to natural noble rot development in the historic Hungarian Tokaj wine region has
Alberto Miceli et al.
International journal of molecular sciences, 21(21) (2020-11-05)
In cognitively normal patients, mild hyperglycemia selectively decreases 18F-Fluorodeoxyglucose (FDG) uptake in the posterior brain, reproducing Alzheimer disease pattern, hampering the diagnostic accuracy of this widely used tool. This phenomenon might involve either a heterogeneous response of glucose metabolism or

Questions

1–3 of 3 Questions  
  1. what all the enzymes it can measure like SOD, CAT, GPX, LPO and GSH activity

    1 answer
    1. This kit is not capable of determining enzymatic activity. The product is intended for use in measuring total antioxidant capacity (TAC) of a given sample as compared to a standard curve based on serial dilutions of Trolox. The kit principal is based on the reduction of Cu2+ to Cu+. Please see the link below to review additional information in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/114/194/mak334bul.pdf

      Note that there are other kit products available for measuring the activity of specific enzymes.

      Helpful?

  2. Can this kit be used with tissue extracts? If yes, how to extract the tissue (liver)?

    1 answer
    1. Yes, this kit is suitable for use with tissue extracts. The tissue preparation recommendations are as follows:
      Homogenize tissue samples on ice in cold 1xPBS.
      (use approximately 0.5 mL PB per 100 mg of tissue)
      Centrifuge at 12,000g for 15 minutes at 4°C.
      Remove the supernatant and keep it on ice.
      Note that the supernatant may be stored in working aliquots at –80°C until ready for testing.

      Helpful?

  3. Is that kit provide instruction for the testing

    1 answer
    1. The product can be found in the 'DOCUMENTATION' section under 'More Documents'. Please use the links below to access the protocol below:
      https://www.sigmaaldrich.com/product/sigma/mak334#product-documentation
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/114/194/mak334bul.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service