T88404
1,3,5-Trithiane
97%
Synonym(s):
sym.-Trithiane, Thioformaldehyde trimer
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C3H6S3
CAS Number:
Molecular Weight:
138.27
Beilstein:
79834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
215-220 °C (dec.) (lit.)
SMILES string
C1SCSCS1
InChI
1S/C3H6S3/c1-4-2-6-3-5-1/h1-3H2
InChI key
LORRLQMLLQLPSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service