Skip to Content
Merck
All Photos(1)

Key Documents

280925

Sigma-Aldrich

2,2-Dimethylpropiophenone

98%

Synonym(s):

tert-Butyl phenyl ketone, Pivalophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC(CH3)3
CAS Number:
Molecular Weight:
162.23
Beilstein:
1906460
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

219-222 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

functional group

ketone
phenyl

SMILES string

CC(C)(C)C(=O)c1ccccc1

InChI

1S/C11H14O/c1-11(2,3)10(12)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

OECPUBRNDKXFDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electrocarboxylation of 2,2-dimethylpropiophenone has been carried out in N-methyl-2-pyrrolidone in a diaphragmless cell equipped with a carbon cathode and an aluminium sacrificial anode.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An unexpected ring carboxylation in the electrocarboxylation of aromatic ketones.
Scialdone ONOFRIO, et al.
Electrochimica Acta, 51(17), 3500-3505 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service