139211
Cyclohexyl phenyl ketone
98%
Synonym(s):
Benzoylcyclohexane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11COC6H5
CAS Number:
Molecular Weight:
188.27
Beilstein:
2046712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
55-57 °C (lit.)
functional group
ketone
phenyl
SMILES string
O=C(C1CCCCC1)c2ccccc2
InChI
1S/C13H16O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12H,2,5-6,9-10H2
InChI key
BMFYCFSWWDXEPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Cyclohexyl phenyl ketone was used to investigate the crosslinked cyclohexyl phenyl compounds under thermal and aquathermal conditions. It was used to examine the effects of molecular changes in the benzophenone molecule on phototoxic behavior.
Biochem/physiol Actions
Cyclohexyl phenyl ketone induces photohaemolysis after exposure to ultraviolet A-rich irradiation in human erythrocytes.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Aqueous organic chemistry. 2. Crosslinked cyclohexyl phenyl compounds.
Siskin M, et al.
Energy and Fuels, 4(5), 482-488 (1990)
Jose Serrano et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(2), 192-208 (2019-03-20)
1. Cyclic phenones are chemicals of interest to the USEPA due to their potential for endocrine disruption to aquatic and terrestrial species.2. Prior to this report, there was very limited information addressing metabolism of cyclic phenones by fish species and
Marianne Placzek et al.
Acta dermato-venereologica, 93(1), 30-32 (2012-09-18)
Benzophenone is a phototoxic compound with absorption maxima in the ultraviolet A (UVA) and ultraviolet B (UVB) range. Many benzophenone derivatives are known to be photosensitizing. On the other hand, 2-hydroxy-4-methoxybenzophenone is used as a photoprotective agent. The aim of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service