Skip to Content
Merck
All Photos(2)

Key Documents

T19550

Sigma-Aldrich

Tetramethylammonium tetrafluoroborate

97%

Synonym(s):

Tetramethylammonium fluoroborate, [Me4N][BF4]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(BF4)
CAS Number:
Molecular Weight:
160.95
Beilstein:
3715404
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

≥300 °C (lit.)

SMILES string

C[N+](C)(C)C.F[B-](F)(F)F

InChI

1S/C4H12N.BF4/c1-5(2,3)4;2-1(3,4)5/h1-4H3;/q+1;-1

InChI key

XWFABLFRYCHILB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ludmila Šimková et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 323, 106895-106895 (2021-01-12)
The molecule of 2,2-dinitroethene-1,1-diamine (FOX-7) is one of the most interesting molecules with multiple redox centres stabilized by push-pull effect. To reveal the detailed mechanism of its electrochemical process radical intermediates formed in the course of its electrochemical reduction in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service