Skip to Content
Merck
All Photos(1)

Key Documents

242144

Sigma-Aldrich

Tetraethylammonium tetrafluoroborate

99%

Synonym(s):

Tetraethylazanium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(BF4)
CAS Number:
Molecular Weight:
217.06
Beilstein:
3917638
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

solid

mp

≥300 °C (lit.)

solubility

acetonitrile: slightly soluble(lit.)

SMILES string

F[B-](F)(F)F.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BF4/c1-5-9(6-2,7-3)8-4;2-1(3,4)5/h5-8H2,1-4H3;/q+1;-1

InChI key

XJRAKUDXACGCHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethylammonium tetrafluoroborate can be used as a supporting electrolyte:
  • In the electrocatalytic reduction of 6-bromo-1-hexene by nickel(I) salen.
  • To synthesize conjugated oligomers via electrochemical polymerization.
  • To fabricate high-performance supercapacitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Andreas Hobisch et al.
Materials (Basel, Switzerland), 13(4) (2020-02-28)
Willow bark is a byproduct from forestry and is obtained at an industrial scale. We upcycled this byproduct in a two-step procedure into sustainable electrode materials for symmetrical supercapacitors using organic electrolytes. The procedure employed precarbonization followed by carbonization using
Szu-Chen Wu et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-04)
Supercapacitors (SCs) are promising for powering mobile devices, electric vehicles and smart power grids due to their fast charge/discharge rate, high power capability and robust cycle stability. Nitrogen-doped porous carbons are great alternatives because they provide pseudocapacitance without losing their
Côme Bodart et al.
ACS applied materials & interfaces, 11(19), 17226-17233 (2019-04-13)
Conducting polymers have been widely explored as coating materials for metal electrodes to improve neural signal recording and stimulation because of their mixed electronic-ionic conduction and biocompatibility. In particular, the conducting polymer poly(3,4-ethylenedioxythiophene) (PEDOT) is one of the best candidates
Ludmila Šimková et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 323, 106895-106895 (2021-01-12)
The molecule of 2,2-dinitroethene-1,1-diamine (FOX-7) is one of the most interesting molecules with multiple redox centres stabilized by push-pull effect. To reveal the detailed mechanism of its electrochemical process radical intermediates formed in the course of its electrochemical reduction in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service