Key Documents
SYNA351420
Synthware distillation adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 14/20, side joint: 14/20, side arm angle: 75 deg
Synonym(s):
Synthware A351420, adapter, sidearm adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
bottom joint: ST/NS14/20
side joint: 14/20
joint: ST/NS top14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A351420
Related Categories
General description
For use as a component in atmospheric or vacuum distillations.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)