Skip to Content
Merck
All Photos(1)

Key Documents

SYNA452440

Synthware inlet/thermometer adapter with one holed rubber adapter

joint: ST/NS 24/40

Synonym(s):

Synthware A452440, thermometer inlet adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

Joint

joint: ST/NS24/40

packaging

pkg of 1 ea

manufacturer/tradename

Synthware A452440

Looking for similar products? Visit Product Comparison Guide

General description

For use with gas inlet tubes or plain stem thermometers. Supplied with one holed rubber adapter.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service