SLW718342
Pyrex® Pestle
rough finish, L 110 mm
Synonym(s):
Pyrex, SciLabware 718342
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
material
borosilicate glass
manufacturer/tradename
718342
SciLabware
L
110 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for crushing solids into powders for easier handling
- Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Available in a rough or glazed finish
- For use with Pyrex Mortars
Features and Benefits
• Manufactured from borosilicate glass 3.3• Available in a rough or glazed finish• Ideal for crushing solids into powders for easier handling• For use with PYREX® Mortars• Pestles and mortars should be ordered separately.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service