Key Documents
Z282081
Sigma-Aldrich
Coors apothecary mortar
O.D. × H 100 mm × 59 mm
About This Item
UNSPSC Code:
41101702
NACRES:
NB.35
material
alumina
manufacturer/tradename
Coorstek 60382
O.D. × H
100 mm × 59 mm
General description
99.5% alumina. Extremely hard and completely nonabsorbent surface. Mortar shaped for easy handling. Outside glazed except for bottom. Order pestle separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)