Skip to Content
Merck
All Photos(1)

Key Documents

QBD-AQ-11918LF

Sigma-Aldrich

AQuora®TX595-NHS Ester

Synonym(s):

AQuora®TX595

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H86N4O22S2
Molecular Weight:
1303.49
UNSPSC Code:
12352108
NACRES:
NA.01

Quality Level

SMILES string

O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNS(C(C=C2S(=O)([O-])=O)=CC=C2C(C3=CC4=C5C(CCCN5CCC4)=C3O6)=C(C=C7CCC8)C6=C9C7=[N+]8CCC9)(=O)=O

General description

AQuora® TX595-NHS Ester is an amine-reactive fluorophore
designed to improve solubility during labeling and of the dye labeled conjugate. As a result, the dye labeled conjugates made with AQuora® TX595-NHS Ester yield enhanced signal and signal-to-noise ratios in fluorescence-based applications including fluorescent western blotting, fluorescence-based microscopy, flow cytometry, and cell-based assays.

This AQuora® TX595-NHS Ester dye is activated with a N-hydroxysuccinimide (NHS) ester group which reacts with free primary amines, such as the amines on the side chain of lysines, forming a stable amide bond.

Legal Information

AQuora is a registered trademark of Quanta BioDesign, Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service