QBD-AQ-11973LF
AQuora®550-NHS Ester
Synonym(s):
1-(31-((2,5-dioxopyrrolidin-1-yl)oxy)-3,31-dioxo-7,10,13,16,19,22,25,28-octaoxa-4-azahentriacontyl)-2-((E)-3-((E)-1-ethyl-3-methyl-5-sulfonato-3-(2,5,8,11-tetraoxatridecan-13-yl)indolin-2-ylidene)prop-1-en-1-yl)-3-methyl
About This Item
Recommended Products
Quality Level
SMILES string
CC(C1=C(N/2CC)C=CC(S(=O)([O-])=O)=C1)(CCOCCOCCOCCOC)C2=C/C=C/C3=[N+](CCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON4C(CCC4=O)=O)=O)=O)C(C=CC(S(=O)([O-])=O)=C5)=C5C3(CCC(NCCOCCOCCOCCOC)=O)C
General description
ThisAQuora® 550-NHS Ester dye is activated with a N-hydroxysuccinimide (NHS) ester group which reacts with free primary amines, such as the amines on the side chain of lysines, forming a stable amide bond.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service