Skip to Content
Merck
All Photos(1)

Key Documents

378151

Sigma-Aldrich

Diglycolyl chloride

95%

Synonym(s):

2,2′-Oxydiacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(CH2COCl)2
CAS Number:
Molecular Weight:
170.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

84-87 °C/2 mmHg (lit.)

density

1.439 g/mL at 25 °C (lit.)

functional group

acyl chloride
ether

SMILES string

ClC(=O)COCC(Cl)=O

InChI

1S/C4H4Cl2O3/c5-3(7)1-9-2-4(6)8/h1-2H2

InChI key

GTZXSBQCNBNWPK-UHFFFAOYSA-N

General description

Diglycolyl chloride (2,2′-Oxydiacetyl chloride) is an acid halide.

Application

Diglycolyl chloride is suitable for use in the synthesis of ply(ether ester). It may be used in the synthesis of:
  • chiral diphenyl substituted polyether-diester compounds
  • morpholine dione analog (IMDNQ)
  • salicylic acid (SA)- based diacids
Diglycolyl chloride may be used in the synthesis of the following compounds:
  • diazadibenzo-18-crown-6 diamide
  • diazadi(tert-butylbenzo)-18-crown-6 diamide
  • surfen derivative

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley L Carbone et al.
Macromolecular rapid communications, 30(12), 1021-1021 (2010-02-18)
Fast-degrading, salicylate-based poly(anhydride-esters) were designed to degrade and release the active component, salicylic acid (SA), within 1 week. The polymer degradation was enhanced by using shorter or oxygen-containing aliphatic chains. A copolymer of diglycolic acid was also made with a
Synthesis and CO2 Solubility Studies of Poly (ether carbonate) s and Poly (ether ester) s Produced by Step Growth Polymerization.
Tan B, et al.
Macromolecules, 38(5), 1691-1698 (2005)
J C Aguilar et al.
Talanta, 54(6), 1195-1204 (2008-10-31)
The ligands 4,7-diaza-2,3,8,9-dibenzo-15-crown-5 (L1), 4,10-diaza-2,3,11,12-dibenzo-18-crown-6 (L2), 4,10-diaza-2,3,11,12-di(4'-tert-butylbenzo)-18-crown-6 (L3) and N,N-di(methylenecarboxyethoxy) 4,10-diaza-2,3,11,12-dibenzo-18-crown-6 (L4) have been prepared. Partition coefficients and acid dissociation constants for these four diazadibenzocrown ether compounds were determined in water-chloroform. Their effectiveness was assessed in solvent extraction of Pb(2+)
Small molecule antagonists of cell-surface heparan sulfate and heparin-protein interactions.
Weiss RJ, et al.
Chemical Science, 6(10), 5984-5993 (2015)
Preparation of chiral diphenyl substituted polyether-diester compounds.
Bradshaw JS, et al.
The Journal of Organic Chemistry, 47(7), 1229-1232 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service