Skip to Content
Merck
All Photos(1)

Key Documents

221015

Sigma-Aldrich

Oxalyl chloride

ReagentPlus®, ≥99%

Synonym(s):

Ethanedioyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOCl
CAS Number:
Molecular Weight:
126.93
Beilstein:
1361988
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.4 (vs air)

Quality Level

vapor pressure

150 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

reaction suitability

reagent type: oxidant

impurities

<10 ppb Heavy metals

color

APHA: 0-150

refractive index

n20/D 1.429 (lit.)

bp

62-65 °C (lit.)

mp

−10-−8 °C (lit.)

density

1.5 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)C(Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6

InChI key

CTSLXHKWHWQRSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxalyl chloride is a commonly used chlorinating reagent that can be prepared by the reaction of oxalic acid and phosphorus pentachloride.

Application

Oxalyl chloride may be used in the following processes:
  • Preparation of Mosher′s acid chloride by reacting with Mosher′s acid in the presence of DMF.
  • Activation of dimethyl sulfoxide for use in the oxidation of long-chain alcohols to carbonyls.
  • Activation of α-keto carboxylic acids and N-heterocyclic carboxylic acids for alkynylation to form ynediones and N-heterocyclic ynones, respectively.
Suitable for the synthesis of acid chlorides used to produce liquid crystals.
Reactant involved in:
  • Synthesis of N-heterocyclic ynones and ynediones, used to activate carboxylic acids
  • Chlorination and halogenation
  • Three-component [3+2] cycloadditions
  • Reactions with organostannanes
  • Synthesis of cyclopentenones
  • Carbonylations, used as a carbonyl synthon

Packaging

The 5g, 25g and 100g units sold in the US are packaged in ampules.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic Syntheses of N-Heterocyclic Ynones and Ynediones by In Situ Activation of Carboxylic Acids with Oxalyl Chloride.
Boersch C, et al.
Angewandte Chemie (International Edition in English), 50(44), 10448-10452 (2011)
A simple method for the microscale preparation of Mosher's acid chloride.
Ward DE and Rhee CK.
Tetrahedron Letters, 32(49), 7165-7166 (1991)
Oxidation of long-chain and related alcohols to carbonyls by dimethyl sulfoxide" activated" by oxalyl chloride.
Mancuso AJ, et al.
The Journal of Organic Chemistry, 43(12), 2480-2482 (1978)
Taisuke Itaya et al.
Chemical & pharmaceutical bulletin, 50(1), 83-86 (2002-02-05)
The relationship between the product patterns and the configurations of 1,2-cycloheptane- and 1,2-cyclooctanediols 9 in the cyclocondensations with oxalyl chloride in the presence of triethylamine at 0 degrees C has been shown analogous to that obtained for 1,2-disubstituted acyclic ethylene
Tsutomu Kimura et al.
Chemical communications (Cambridge, England), (32)(32), 4077-4079 (2005-08-11)
Reaction of diastereomerically pure phosphinoselenoic acid salts with oxalyl chloride leads to enantiomerically pure P-chiral phosphinoselenoic chlorides with inversion of configuration at phosphorus; one of these chlorides is converted to a phosphinoselenothioic acid salt with a high degree of enantioselectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service