Skip to Content
Merck
All Photos(1)

Key Documents

A38800

Sigma-Aldrich

2-Aminoanthracene

96%

Synonym(s):

2-Anthramine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Beilstein:
2209414
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

powder

mp

238-241 °C (lit.)

SMILES string

Nc1ccc2cc3ccccc3cc2c1

InChI

1S/C14H11N/c15-14-6-5-12-7-10-3-1-2-4-11(10)8-13(12)9-14/h1-9H,15H2

InChI key

YCSBALJAGZKWFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminoanthracene can be used as a reactant to prepare:
  • Steroid derived naphthoquinoline asphaltene compounds via multicomponent cyclocondensation reaction with 5-α-cholestan-3-one and aromatic aldehydes.
  • Naphtho[2,3- f ]quinoline derivatives by reacting with aromatic aldehyde and acetone or acetophenone catalyzed by iodine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steroid-Derived Naphthoquinoline Asphaltene Model Compounds: Hydriodic Acid Is the Active Catalyst in I2-Promoted Multicomponent Cyclocondensation Reactions
Schulze M, et al.
Organic Letters, 17(23), 5930-5933 (2015)
An efficient method for the synthesis of naphthoquinoline derivatives catalyzed by iodine
Wang W, et al.
Heterocyclic Communications, 18(1), 17-21 (2012)
Junichiro Saito et al.
Mutation research, 609(1), 68-73 (2006-08-04)
Magnolol, a component of the bark of Magnolia obovata, has been reported to possess various biological activities, such as anti-carcinogenicity, anti-promotion activity and anti-oxidative activity. These findings suggest potential for this compound in cancer chemoprevention. Interestingly, there have been no
A Di Sotto et al.
Journal of ethnopharmacology, 127(3), 731-736 (2009-12-09)
The aerial parts of Sisymbrium officinale Scop. are commonly used to treat airway ailments, moreover in antiquity the herbal drug was reputed to possess anticancer properties. The results obtained in present work support the traditional use and the properties ascribed
Rodrigo Juliano Oliveira et al.
Toxicology in vitro : an international journal published in association with BIBRA, 20(7), 1225-1233 (2006-05-24)
Due to the need to identify new antimutagenic agents and to determine their mechanism of action, the present study examined the mechanism of action of the beta-glucan with regard to antimutagenicity using the micronucleus assay in CHO-k1 and HTC cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service