Skip to Content
Merck
All Photos(1)

Key Documents

103594

Sigma-Aldrich

1-Nitronaphthalene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
1867714
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

bp

304 °C (lit.)

mp

53-57 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble
carbon disulfide: freely soluble
chloroform: soluble
diethyl ether: freely soluble

density

1.223 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc2ccccc12

InChI

1S/C10H7NO2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

RJKGJBPXVHTNJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Nitronaphthalene is a mutagenic nitroaromatic compound present in diesel exhaust and it causes acute liver and lung toxicity in rodents[1]. 1-Nitronaphthalene is a cytochrome P450-bioactivated, nonciliated bronchiolar epithelial (Clara) cell cytotoxicant [2].

Application

1-Nitronaphthalene was used to study excited state dynamics of 1-nitropthalene in nonpolar, aprotic and protic solvents[3].

Biochem/physiol Actions

1-Nitronaphthalene binds covalently to protein and forms adducts in the rat airway epithelium in vivo[4].

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christian Reichardt et al.
The Journal of chemical physics, 131(22), 224518-224518 (2009-12-17)
The electronic energy relaxation of 1-nitronaphthalene was studied in nonpolar, aprotic, and protic solvents in the time window from femtoseconds to microseconds. Excitation at 340 or 360 nm populates the Franck-Condon S(1)(pipi( *)) state, which is proposed to bifurcate into
J M Sauer et al.
Toxicology letters, 90(1), 19-27 (1997-01-15)
1-Nitronaphthalene is a mutagenic particulate of diesel exhaust which causes acute liver and lung toxicity in rodents. The studies presented here describe morphological changes in the lung and liver at several time intervals following a single injection of 1-nitronaphthalene (100
K el-Bayoumy et al.
Carcinogenesis, 6(4), 505-507 (1985-04-01)
A method was developed for the analysis of 1-nitronaphthalene, 1-nitropyrene, and 6-nitrochrysene in mainstream cigarette smoke, using [14C]1-nitronaphthalene, [14C]1-nitropyrene, and [14C]6-nitrochrysene as tracers and internal standards. Cigarette smoke condensate was collected and the appropriate fractions containing the labelled standards were
Gregory L Baker et al.
Toxicological sciences : an official journal of the Society of Toxicology, 77(1), 135-141 (2003-11-06)
The mechanisms of toxicant-mediated lung injury and repair are influenced by the considerable spatial heterogeneity that exists within the conducting airways of the lungs. As a result of this heterogeneity, significant differences and similarities in gene expression are observed throughout
Ching Yu Lin et al.
Proteomics, 6(3), 972-982 (2006-02-03)
Naphthalene and 1-nitronaphthalene are ambient air pollutants, which undergo P450-dependent bioactivation in the lung. Reactive metabolites of naphthalene and 1-nitronaphthalene covalently bind to proteins, and the formation of covalent adducts correlates with airway epithelial cell injury in rodent models. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service