Skip to Content
Merck
All Photos(1)

Key Documents

H2805

Sigma-Aldrich

1-Heptanol

98%

Synonym(s):

Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
Beilstein:
1731686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.5 mmHg ( 20 °C)

Quality Level

Assay

98%

form

liquid

autoignition temp.

662 °F

refractive index

n20/D 1.424 (lit.)

bp

176 °C (lit.)

mp

−36 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCO

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Heptanol can be used:
  • As a co-solvent in binary and ternary systems to estimate the liquid-liquid interaction parameters.
  • In the synthesis of heptyl oleate ester from oleic acid.
  • As a reducing agent in the synthesis of copper nanoparticles via hydroxyl ion-assisted alcohol reduction of copper salts in the presence of oleylamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid- liquid equilibria of the ternary system water+ acetic acid+ 1-heptanol
Aljimaz AS, et al.
Journal of Chemical and Engineering Data, 45(2), 301-303 (2000)
Copper nanoparticles synthesized by hydroxyl ion assisted alcohol reduction for conducting ink
Huaman JLC, et al.
Journal of Materials Chemistry, 21(20), 7062-7069 (2011)
Screening and catalytic activity in organic synthesis of novel fungal and yeast lipases
Cardenas F, et al.
Journal of Molecular Catalysis. B, Enzymatic, 14(4-6), 111-123 (2001)
Density and viscosity of binary mixtures of {1-butyl-3-methylimidazolium thiocyanate+ 1-heptanol, 1-octanol, 1-nonanol, or 1-decanol}
Domanska U and Krolikowska M
Journal of Chemical and Engineering Data, 55(9), 2994-3004 (2010)
Nannan Ye et al.
Lab on a chip, 11(6), 1096-1101 (2011-02-05)
Transport across gap junction channels (GJCs) between neighboring cells is critical to synchronizing cell's electrical and metabolic activities and maintaining cell homeostasis. Here we present a non-invasive microfluidic method to measure molecular diffusion across GJCs in multiple 1D cell arrays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service