Skip to Content
Merck
All Photos(2)

Key Documents

K401

Sigma-Aldrich

2-Ketobutyric acid

97%

Synonym(s):

2-Oxobutanoic acid, 2-Oxobutyric acid, α-Ketobutyric acid, Propionylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOOH
CAS Number:
Molecular Weight:
102.09
Beilstein:
1700514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

bp

84 °C/20 mmHg (lit.)

mp

30-34 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(=O)C(O)=O

InChI

1S/C4H6O3/c1-2-3(5)4(6)7/h2H2,1H3,(H,6,7)

InChI key

TYEYBOSBBBHJIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ketobutyric acid can be used as a reactant to synthesize:
  • (S)-2-Aminobutyric acid via enzyme-catalyzed amination reaction.
  • L-Isoleucine by living cell reaction in the presence of ethanol.
  • 3-Ethyl-2-quinoxalinol from 1,2-diaminobenzene in the presence of sulfuric acid.
  • 3-Ethylbenzo[g]quinoxalin-2(1H)-one using 2,3-diaminonaphthalene in the presence of biocatalyst.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of potential chemotherapic quinoxalinone derivatives by biocatalysis or microwave-assisted Hinsberg reaction
Gris J, et al.
Tetrahedron Letters, 49, 1053-1056 (2008)
Fabrication of a microcapsule extinguishing agent with a core-shell structure for lithium-ion battery fire safety
Zhang W, et al.
Materials Advances, 2, 4634-4642 (2021)
Synthesis of 2-(Quinoxalin-2-ylamino-benzotriazolyl) Pentanedioic Derivatives as Potential Anti-Folate Agents
Briguglio I, et al.
Journal of Heterocyclic Chemistry, 53, 1721-1737 (2016)
Thomas Svoboda et al.
Frontiers in plant science, 10, 1072-1072 (2019-09-26)
Fusarium graminearum is a plant pathogenic fungus which is able to infect wheat and other economically important cereal crop species. The role of ethylene in the interaction with host plants is unclear and controversial. We have analyzed the inventory of
Nora Weber et al.
Microbial cell factories, 16(1), 51-51 (2017-03-25)
Saccharomyces cerevisiae (baker's yeast) has great potential as a whole-cell biocatalyst for multistep synthesis of various organic molecules. To date, however, few examples exist in the literature of the successful biosynthetic production of chemical compounds, in yeast, that do not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service