464422
2-Ketobutyric acid sodium salt hydrate
≥95%
Synonym(s):
Sodium 2-Ketobutyrate Hydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
≥95%
form
powder
impurities
<5% corresponding acid
mp
210 °C (dec.) (lit.)
functional group
ketone
storage temp.
2-8°C
SMILES string
O.[Na+].CCC(=O)C([O-])=O
InChI
1S/C4H6O3.Na.H2O/c1-2-3(5)4(6)7;;/h2H2,1H3,(H,6,7);;1H2/q;+1;/p-1
InChI key
PVLJVKQSCAHPPR-UHFFFAOYSA-M
Related Categories
General description
2-Ketobutyric acid sodium salt hydrate is widely used in organic synthesis as a valuable building block for the preparation bioactive compounds. It is also used as a precursor to synthesize alanine, which is an important amino acid.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols for analyzing metabolic derangements caused by increased NADH/NAD+ ratio in cell lines and in mice
STAR protocols, 3, 101120-101120 (2022)
Selective Alanine Transporter Utilization Creates a Targetable Metabolic Niche in Pancreatic Cancer.
Cancer discovery, 10(7), 1018-1037 (2020-04-29)
Pancreatic ductal adenocarcinoma (PDAC) evolves a complex microenvironment comprised of multiple cell types, including pancreatic stellate cells (PSC). Previous studies have demonstrated that stromal supply of alanine, lipids, and nucleotides supports the metabolism, growth, and therapeutic resistance of PDAC. Here
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service