Skip to Content
Merck
All Photos(2)

Key Documents

75940

Sigma-Aldrich

2-Oxooctanoic acid

≥99.0% (T)

Synonym(s):

α-Ketooctanoic acid, 2-Oxocaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COCOOH
CAS Number:
Molecular Weight:
158.19
Beilstein:
1757862
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

flakes

bp

82 °C/0.05 mmHg

mp

33-36 °C

functional group

carboxylic acid
ketone

storage temp.

2-8°C

SMILES string

CCCCCCC(=O)C(O)=O

InChI

1S/C8H14O3/c1-2-3-4-5-6-7(9)8(10)11/h2-6H2,1H3,(H,10,11)

InChI key

GPPUPQFYDYLTIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huifang Xu et al.
Journal of colloid and interface science, 509, 265-274 (2017-09-16)
Single-tailed short-chain alkyl keto-acids/salts, a class of fatty acid/salt derivatives, such as sodium 2-ketooctanate (KOCOONa), are a kind of weakly acid/salt type amphiphiles and plausible prebiotic molecules, and the current understanding of their aggregation behavior in aqueous solutions is still
Sarah A Almahboub et al.
Applied microbiology and biotechnology, 102(2), 789-799 (2017-11-28)
Terminal modification of peptides is frequently used to improve their hydrophobicity. While N-terminal modification with fatty acids (lipidation) has been reported previously, C-terminal lipidation is limited as it requires the use of linkers. Here we report the use of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service