Key Documents
HAM203230
Hamilton® SaltLine reagent syringe
model, 1750 TLL-SAL, no needle, volume 500 μL
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
Pricing and availability is not currently available.
material
Gastight syringe
feature
autoclavable: no
model 1750 TLL-SAL, no needle
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
700 psig pressure
volume
500 μL
General description
The SaltLine syringes are utilized in applications with high salt concentrations. They have the same PTFE Luer Lock termination but the plunger tip is made from Ultra High Molecular Weight Poly Ethylene (UHMWPE). The UHMWPE plunger tip resists damage that can be caused when salt crystal precipitate from the salt solution over time.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)