Key Documents
20720
Hamilton® syringe, 1000 series GASTIGHT®, luer tip
1001LT, volume 1 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Pricing and availability is not currently available.
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81301
needle L
, n/a
volume
1 mL
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
207222072120723
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
needle size
(not included)
needle size
(not included)
needle size
(not included)
needle size
(not included)
feature
without needle Luer tip
feature
without needle Luer tip
feature
without needle Luer tip
feature
without needle Luer tip
manufacturer/tradename
Hamilton 81301
manufacturer/tradename
Hamilton 81501
manufacturer/tradename
Hamilton 81401
manufacturer/tradename
Hamilton 81601
needle L
, n/a
needle L
, n/a
needle L
, n/a
needle L
, n/a
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)