Skip to Content
Merck
All Photos(2)

Key Documents

GF97195595

Indium

foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.8%

Synonym(s):

Indium, IN000050

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

≥99.8%

form

foil

manufacturer/tradename

Goodfellow 971-955-95

resistivity

8.37 μΩ-cm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Schulz et al.
Medical physics, 4(1), 42-45 (1977-01-01)
A sensitive and relatively simple method for obtaining an index of the spectral quality of x rays in the 4-15-MV range is presented. The method is based upon the simultaneous production of 115mIn by inelastic scattering of x rays by
Evaluation of the repair of mechanical furcation perforations using amalgam, gutta-percha, or indium foil.
R Aguirre et al.
Journal of endodontics, 12(6), 249-256 (1986-06-01)
Loïc Bertrand et al.
The Analyst, 138(16), 4463-4469 (2013-04-05)
In order to fully characterize the zinc white artists' pigment (ZnO), much used since the mid-nineteenth century, three samples collected in the early 20th century were studied using a combination of synchrotron and macroscopic photoluminescence spectroscopy and imaging. An improved
M K Loken et al.
Clinical nuclear medicine, 10(12), 902-911 (1985-12-01)
Following the introduction of In-111 oxine as a label for blood cells by McAffee and Thakur in 1976, these procedures have become increasingly important in the practice of nuclear medicine. Of particular interest are studies involving the use of labeled
New generation of palladium indium-silver dental cast alloys: a review.
M L Malhotra
The Alpha omegan, 88(1), 28-30 (1995-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service