Skip to Content
Merck
Indium wire reel, 5m, diameter 1.5mm, as drawn, 99.999%

Indium

Indium wire reel, 5m, diameter 1.5mm, as drawn, 99.999%
Synonym(s):
Indium
Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11