Merck
Clinical use of indium-111 labeled blood products.
Clinical nuclear medicine (1985-12-01)
M K Loken, M E Clay, R T Carpenter, R J Boudreau, J J McCullough
ABSTRACT
Following the introduction of In-111 oxine as a label for blood cells by McAffee and Thakur in 1976, these procedures have become increasingly important in the practice of nuclear medicine. Of particular interest are studies involving the use of labeled leukocytes for the detection of focal infection. The clinical utility of labeled platelets is less well developed, although the use of platelets to detect the formation of thrombi in blood vessels and on vascular grafts and prostheses is gaining prominence. This report summarizes the techniques presently employed at the University of Minnesota for the labeling of blood products, and their clinical use. Consideration also is given to the desired expertise and cost factors involved in the labeling of leukocytes and platelets.
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Indium, foil, not light tested, 25x25mm, thickness 0.007mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 25x25mm, thickness 0.008mm, temporary acrylic support, 99.8%
Indium, wire reel, 5m, diameter 1.0mm, as drawn, 99.999%
Indium, wire reel, 0.5m, diameter 1.5mm, as drawn, 99.999%
Indium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.04mm, 99.8%
Indium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 100%
Indium, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.8%
Indium, foil, not light tested, 50x50mm, thickness 0.009mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.8%
Indium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 100%