Skip to Content
Merck
All Photos(1)

Key Documents

D72204

Sigma-Aldrich

1,3-Dichloropropane

99%

Synonym(s):

Trimethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3Cl
CAS Number:
Molecular Weight:
112.99
Beilstein:
505960
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

120-122 °C (lit.)

mp

−99 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

ClCCCCl

InChI

1S/C3H6Cl2/c4-2-1-3-5/h1-3H2

InChI key

YHRUOJUYPBUZOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rodriguez-Arnaiz
Environmental and molecular mutagenesis, 31(4), 390-401 (1998-07-08)
Biotransformation of several structurally related 2B compounds to reactive metabolites was evaluated in the somatic w/w+ assay of Drosophila melanogaster. Chemicals tested were the dichlorinated alkanes dichloromethane (DCM), 1,2-dichloroethane (DCE), and 1,3-dichloropropane (DCP); the thiouracil derivatives 5-methyl, 2-thiouracil (5M2TU), 6-methyl
Stefaan De Neve et al.
Journal of environmental quality, 33(5), 1647-1652 (2004-09-10)
The effect of soil fumigation on N mineralization and nitrification needs to be better quantified to optimize N fertilizer advice and predict NO(-)(3) concentrations in crops and NO(-)(3) leaching risks. Seven soils representing a range in soil texture and organic
S Shin et al.
Chemico-biological interactions, 97(3), 229-238 (1995-08-18)
Treatment of a lysosome-rich fraction from liver with 2-chloroethylethyl sulfide resulted in a dose-dependent release of arylsulfatase. The inclusion of Ca2+ enhanced the enzyme release by approximately 2.3-fold. The enhancing effect of Ca2+, showing an EC50 value of 30 mM
1,3-Dichloropropene (technical grade).
Report on carcinogens : carcinogen profiles, 10, 91-93 (2004-08-25)
Biological effect monitoring.
C T Evelo et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 15, 268-277 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service