Skip to Content
Merck
All Photos(1)

Key Documents

02577

Supelco

1,2-Dichloropropane

analytical standard

Synonym(s):

Propylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCH2Cl
CAS Number:
Molecular Weight:
112.99
Beilstein:
1718880
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.89 (vs air)

vapor pressure

40 mmHg ( 19.4 °C)

Assay

≥99.5% (GC)

autoignition temp.

1035 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

14.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.439 (lit.)
n20/D 1.439

bp

95-96 °C (lit.)

mp

−100 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Cl)CCl

InChI

1S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3

InChI key

KNKRKFALVUDBJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloropropane (1,2-D), is a colorless watery liquid with a sweet odor. It sinks in water and produces irritating vapor. It is a short-chain chlorinated aliphatic compound, commonly used as fumigant and as insecticide. It is a widespread groundwater contaminant. Anaerobic bacteria have the capacity to reductively dechlorinate it to propene.

Application

1,2-Dichloropropane may be used as an analytical reference standard for the determination of the analyte in wastewater, marine water, drinking water and air samples by gas chromatography (GC) technique.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous quantification of polar and non-polar volatile organic compounds in water samples by direct aqueous injection-gas chromatography/mass spectrometry
Aeppli C, et al.
Journal of Chromatography A, 1181(1-2), 116-124 (2008)
A sequential analysis for volatile organics in water by purge-and-trap capillary column gas chromatography with photoionization and electrolytic conductivity detectors in series
Ho JSY
Journal of Chromatographic Science, 27(2), 91-98 (1989)
Continuous determination of volatile organic compounds in the atmosphere by an automated gas chromatographic system
Yamamoto N, et al.
Journal of Chromatography A, 819(1-2), 177-186 (1998)
Kovi Bessoff et al.
Antimicrobial agents and chemotherapy, 57(4), 1804-1814 (2013-02-06)
Cryptosporidiosis, a diarrheal disease usually caused by Cryptosporidium parvum or Cryptosporidium hominis in humans, can result in fulminant diarrhea and death in AIDS patients and chronic infection and stunting in children. Nitazoxanide, the current standard of care, has limited efficacy
Evaluation of purge-and-trap-high-resolution gas chromatography-mass spectrometry for the determination of 27 volatile organic compounds in marine water at the ng l-1 concentration level
Huybrechts T, et al.
Journal of Chromatography A, 893(2), 367-382 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service